P5630
3,4-Dihydroxybenzoic acid
Synonym(s):
Protocatechuic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
1448841
EC Number:
MDL number:
UNSPSC Code:
12352100
Recommended Products
mp
197-200 °C (dec.) (lit.)
SMILES string
OC(=O)c1ccc(O)c(O)c1
InChI
1S/C7H6O4/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,8-9H,(H,10,11)
InChI key
YQUVCSBJEUQKSH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Chemopreventive in several animal models of carcinogenesis. Blocks cell proliferation in the post-initiation phase.
Other Notes
This product has been replaced by 37580-ALDRICH | 3,4-Dihydroxybenzoic acid ≥97.0% (T)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Frédéric Martin et al.
Journal of natural products, 72(5), 852-856 (2009-04-14)
Extracts from several plants of the family Bignoniaceae from Panama were submitted to a rapid DPPH TLC test for the detection of radical-scavenging activity. The MeOH extract of the stems of Jacaranda caucana, a tree that grows from Costa Rica
R Masella et al.
Current medicinal chemistry, 19(18), 2901-2917 (2012-04-24)
Epidemiological evidence has shown that a high dietary intake of vegetables and fruit rich in polyphenols is associated with a reduction of cancer incidence and mortality from coronary heart disease. The healthy effects associated with polyphenol consumption have made the
Paola Vitaglione et al.
The Journal of nutrition, 137(9), 2043-2048 (2007-08-22)
The metabolic fate of dietary anthocyanins (ACN) has not been fully clarified in humans. In all previous studies, the proportion of total ACN absorbed and excreted in urine was <1% intake. This study aimed to elucidate the human metabolism of
Lei Wang et al.
Environment international, 59, 27-32 (2013-06-12)
Human exposure to p-hydroxybenzoic acid esters (parabens) is a concern, owing to adverse health effects of these compounds. Parabens are metabolized and eliminated from the human bodies within a few hours of exposure. In this study, for the first time
J Kristinsson et al.
Pharmacology & toxicology, 74(3), 174-180 (1994-03-01)
The urinary metabolites of orally administered mebeverine hydrochloride (270 mg) were studied in five healthy volunteers with the aid of gas chromatography/mass spectrometry. Mebeverine, which is an ester of veratric acid and 4-(ethyl-[2-(4-methoxyphenyl)-1-methylethyl]amino)butan-1-ol, was completely hydrolysed to the corresponding acid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service