P0426
Hexaethylene glycol monohexadecyl ether
Synonym(s):
C16E6, Hexadecylhexaglycol
About This Item
description
non-ionic
SMILES string
CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCO
InChI
1S/C28H58O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17-30-19-21-32-23-25-34-27-28-35-26-24-33-22-20-31-18-16-29/h29H,2-28H2,1H3
InChI key
UOFCAQWHTQFNHS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service