Skip to Content
Merck
All Photos(1)

Key Documents

O5126

Sigma-Aldrich

Oligomycin B

~95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C45H72O12
CAS Number:
Molecular Weight:
841.08
Beilstein:
5705502
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

~95% (HPLC)

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

[H][C@@]12CC[C@H](CC)\C=C\C=C\C[C@H](C)[C@@H](O)[C@](C)(O)C(=O)[C@H](C)[C@@H](O)[C@H](C)C(=O)[C@H](C)[C@@H](O)[C@H](C)\C=C\C(=O)O[C@@]([H])([C@H]1C)[C@@H](C)[C@]3(O2)O[C@]([H])(C[C@H](C)O)[C@H](C)CC3=O

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service