Skip to Content
Merck
All Photos(1)

Key Documents

N7001

Sigma-Aldrich

2-Nitro-4-carboxyphenyl N,N-diphenylcarbamate

Synonym(s):

NCDC

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H14N2O6
CAS Number:
Molecular Weight:
378.33
MDL number:
UNSPSC Code:
12352202

SMILES string

OC(=O)c1ccc(OC(=O)N(c2ccccc2)c3ccccc3)c(c1)N(=O)=O

Application

Can be used for the determination of chymotrypsin; liberates 4-hydroxy-3-nitrobenzoic acid (also available from Sigma).

Biochem/physiol Actions

Specific chromogenic inactivator of chymotrypsin. Inhibits anti-IgE-mediated histamine release and inositol trisphosphate (IP3) production in mast cells, possibly by a mechanism involving inhibition of phospholipase C (PLC).[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Takei et al.
Biochemical and biophysical research communications, 181(3), 1313-1322 (1991-12-31)
NCDC dose-dependently inhibited histamine release from rat peritoneal mast cells induced by anti-IgE. Moreover, NCDC inhibited Ca(2+)-mobilization from intracellular Ca(2+)-stores as well as histamine release in mast cells activated by anti IgE, the effect on both of these phenomena being
THE UTILIZATION OF A SPECIFIC CHROMOGENIC INACTIVATOR IN AN "ALL OR NONE" ASSAY FOR CHYMOTRYPSIN.
B F ERLANGER et al.
Biochemistry, 3, 346-349 (1964-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service