Skip to Content
Merck
All Photos(1)

Key Documents

N6523

Sigma-Aldrich

Anti-Nephronophthisis (NPH) (pic) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
2.780,00 kr.

2.780,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
250 μG
2.780,00 kr.

About This Item

UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.41

2.780,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 90 kDa

species reactivity

mouse

technique(s)

immunoprecipitation (IP): 10-20 μg/mL
western blot: 2-10 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12Cc3c[nH]c4cccc(C1=C[C@H](CN2)C(O)=O)c34

InChI key

VGLIYJAMHMSYBY-NOZJJQNGSA-N

Gene Information

mouse ... Nphp1(53885)

Related Categories

General description

Nephronophthisis 1 homolog (Nph) is highly expressed in mouse testis.

Immunogen

GST fusion protein corresponding to amino acids 612-691 of mouse NPH.

Biochem/physiol Actions

Studies have shown that male mice with a mutant nephronophthisis 1 homolog (Nph) protein turn out to be infertile.

Physical form

Solution in phosphate buffered saline containing 0.08% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeted disruption of Nphp1 causes male infertility due to defects in the later steps of sperm morphogenesis in mice.
Jiang ST
Human Molecular Genetics, 17(21), 3368-3379 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service