Skip to Content
Merck
All Photos(2)

Key Documents

N4540

Sigma-Aldrich

NA2F Glycan

from Porcine, ≥90% (HPLC)

Synonym(s):

(Gal)2(GlcNAc)2(Man)3(GlcNAc)2Fuc, Asialo, galactosylated, biantennary N-glycan, core fucosylated, Mannotriose-[fucosyl-di-(N-acetylglucosamine)], bis(galactosyl-N-acetylglucosaminyl)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
4.270,00 kr.

4.270,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
4.270,00 kr.

About This Item

Empirical Formula (Hill Notation):
C68H114N4O50
CAS Number:
Molecular Weight:
1787.63
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

4.270,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

Porcine

Quality Level

Assay

≥90% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC1OC(OCC(O)C(OC2OC(CO)C(OC3OC(COC4OC(CO)C(O)C(O)C4OC5OC(CO)C(OC6OC(CO)C(O)C(O)C6O)C(O)C5NC(C)=O)C(O)C(OC7OC(CO)C(O)C(O)C7OC8OC(CO)C(OC9OC(CO)C(O)C(O)C9O)C(O)C8NC(C)=O)C3O)C(O)C2NC(C)=O)C(O)C(NC(C)=O)C=O)C(O)C(O)C1O

InChI

1S/C68H114N4O50/c1-16-34(86)44(96)49(101)63(107-16)105-14-22(85)53(35(87)21(6-73)69-17(2)81)116-60-31(70-18(3)82)41(93)56(29(13-80)112-60)119-66-52(104)57(120-68-59(48(100)39(91)26(10-77)111-68)122-62-33(72-20(5)84)43(95)55(28(12-79)114-62)118-65-51(103)46(98)37(89)24(8-75)109-65)40(92)30(115-66)15-106-67-58(47(99)38(90)25(9-76)110-67)121-61-32(71-19(4)83)42(94)54(27(11-78)113-61)117-64-50(102)45(97)36(88)23(7-74)108-64/h6,16,21-68,74-80,85-104H,7-15H2,1-5H3,(H,69,81)(H,70,82)(H,71,83)(H,72,84)

InChI key

RSHPVGDNLRQFMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

NA2F N-glycan is a fucosylated biantennary oligosaccharide (FBO) that may be found in the Fc region of IgG. In a study of glycans isolated from a recombinant monoclonal IgG1, high-mannose oligosaccharides were reported to be cleared at a faster rate than FBOs. Changes to NA2F have been associated with health in human aging.

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service