Skip to Content
Merck
All Photos(1)

Key Documents

N4411

Sigma-Aldrich

NU6027

≥98% (HPLC)

Synonym(s):

4-Cyclohexylmethoxy-2,6-diamino-5-nitrosopyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N5O2
CAS Number:
Molecular Weight:
251.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

lavender

mp

252.5-253.7 °C (lit.)

solubility

DMSO: 15 mg/mL

storage temp.

2-8°C

SMILES string

O=NC1=C(N)N=C(N)N=C1OCC2CCCCC2

InChI

1S/C11H17N5O2/c12-9-8(16-17)10(15-11(13)14-9)18-6-7-4-2-1-3-5-7/h7H,1-6H2,(H4,12,13,14,15)

InChI key

DGWXOLHKVGDQLN-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

NU6027 is a potent inhibitor of ATR signalling
Selective cyclin-dependent kinase-2 (CDK2) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Peasland et al.
British journal of cancer, 105(3), 372-381 (2011-07-07)
The ataxia telangiectasia mutated and Rad3-related kinase (ATR) has a key role in the signalling of stalled replication forks and DNA damage to cell cycle checkpoints and DNA repair. It has long been recognised as an important target for cancer
Rebeka Sultana et al.
PloS one, 8(2), e57098-e57098 (2013-03-02)
Ataxia telangiectasia mutated and Rad3 Related (ATR) protein kinase is a key sensor of single-stranded DNA associated with stalled replication forks and repair intermediates generated during DNA repair. XRCC1 is a critical enzyme in single strand break repair and base
Sarah Akbib et al.
PloS one, 14(2), e0212210-e0212210 (2019-02-20)
Cell therapy for diabetes could benefit from the identification of small-molecule compounds that increase the number of functional pancreatic beta cells. Using a newly developed screening assay, we previously identified glucocorticoids as potent stimulators of human and rat beta cell
Jie Chen et al.
DNA repair, 73, 55-63 (2018-11-25)
The ataxia telangiectasia-mutated and Rad3-related (ATR) serine/threonine kinase plays a central role in the repair of replication-associated DNA damage, the maintenance of S and G2/M-phase genomic stability, and the promotion of faithful mitotic chromosomal segregation. A number of stimuli activate
Benjamin A Hilton et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 31(9), 3882-3893 (2017-05-19)
Hutchinson-Gilford progeria syndrome (HGPS) is a rare genetic disorder that is caused by a point mutation in the LMNA gene, resulting in production of a truncated farnesylated-prelamin A protein (progerin). We previously reported that XPA mislocalized to the progerin-induced DNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service