Skip to Content
Merck
All Photos(1)

Key Documents

N1134

Sigma-Aldrich

Se-(p-Nitrobenzyl)-6-selenoinosine

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H17SeN5O6
CAS Number:
Molecular Weight:
466.31
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥95%

form

solid

storage temp.

−20°C

SMILES string

OCC1OC(C(O)C1O)n2cnc3c([Se]Cc4ccc(cc4)N(=O)=O)ncnc23

InChI

1S/C17H17N5O6Se/c23-5-11-13(24)14(25)17(28-11)21-8-20-12-15(21)18-7-19-16(12)29-6-9-1-3-10(4-2-9)22(26)27/h1-4,7-8,11,13-14,17,23-25H,5-6H2

InChI key

XCNANXPIYMOXKW-UHFFFAOYSA-N

Biochem/physiol Actions

Se-(p-nitrobenzyl)-6-selenoinosine is a selenium containing inosine derivative that may have activities similar to S-(p-nitrobenzyl)-6-thioinosine (NBTI), an inhibitor of adenosine equilibrative transporters.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Timóteo et al.
Neurochemistry international, 52(4-5), 834-845 (2007-11-21)
The influence of nerve stimulation pattern on transmitter release inhibition by L-citrulline, the co-product of NO biosynthesis by nitric oxide synthase (NOS), was studied in the rat phrenic nerve-hemidiaphragm. We also investigated the putative interactions between NOS pathway and the
Alexandre dos Santos-Rodrigues et al.
Biochemical and biophysical research communications, 414(1), 175-180 (2011-09-29)
Adenosine is an important modulator of neuronal survival and differentiation in the CNS. Our previous work showed that nucleoside transporters (NTs) are present in cultures of chick retinal cells, but little is known about the mechanisms regulating adenosine transport in
Sílvia V Conde et al.
Journal of neurochemistry, 89(5), 1148-1156 (2004-05-19)
The effect of hypoxia on the release of adenosine was studied in vitro in the rat whole carotid body (CB) and compared with the effect of hypoxia (2%, 5% and 10% O(2)) on adenosine concentrations in superior cervical ganglia (SCG)
Laura Oliveira et al.
Neuroscience letters, 459(3), 127-131 (2009-05-12)
At the rat motor nerve terminals, activation of muscarinic M(1) receptors negatively modulates the activity of inhibitory muscarinic M(2) receptors. The present work was designed to investigate if the negative crosstalk between muscarinic M(1) and M(2) autoreceptors involved endogenous adenosine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service