Skip to Content
Merck
All Photos(1)

Documents

N0376

Sigma-Aldrich

1-Naphthyl propionate

Synonym(s):

Propionic acid 1-naphthyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12O2
CAS Number:
Molecular Weight:
200.23
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CCC(=O)Oc1cccc2ccccc12

InChI

1S/C13H12O2/c1-2-13(14)15-12-9-5-7-10-6-3-4-8-11(10)12/h3-9H,2H2,1H3

InChI key

FRXPDEZCWCPLIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

Esterase substrate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P C Wagaman et al.
Journal of clinical microbiology, 27(5), 832-836 (1989-05-01)
A variety of chemically defined compounds were tested to characterize the substrate specificity of the influenza C virus esterase and to determine whether a substrate could be found that would be useful in an assay to detect the virus. Two
Marsha M Wheeler et al.
Archives of insect biochemistry and physiology, 73(1), 30-48 (2009-10-06)
Four esterase genes and general esterase activity were investigated in the gut of the termite Reticulitermes flavipes. Two genes (RfEst1 and RfEst2) share significant translated identity with a number of insect JH esterases. The two remaining genes (RfEst3 and RfEst4)
Yiou Pan et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 152(3), 266-270 (2008-12-27)
Levels of insecticide resistance, carboxylesterase activity, carboxylesterase expression, and the cDNA sequence of carboxylesterase gene were investigated in malathion resistant and susceptible strains of cotton aphids, Aphis gossypii (Glover). The resistant strain (MRR) exhibited 80.6-fold resistance to malathion compared to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service