Key Documents
MTCV5809R
MTC Bio Cap Inserts for Cryogenic Vials
red
Synonym(s):
PCR strip opener, cryo storage identification, cryo vial marker, tuber holder
About This Item
UNSPSC Code:
41122500
NACRES:
NB.11
Pricing and availability is not currently available.
material
polypropylene
red
packaging
pack of 500 ea
manufacturer/tradename
MTC Bio V5809-R
General description
Cap inserts for cryogenic vials, red, 500/pk
Easily color-code your cryogenic vials for quick visual identification in the lab, for use at temperatures as low as -196°C.
Features and Benefits
  • For temperatures as low as -196°C
Legal Information
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)