Skip to Content
Merck
All Photos(1)

Key Documents

MAK493

Sigma-Aldrich

Isocitrate Dehydrogenase (IDH) Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
3.460,00 kr.

3.460,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
3.460,00 kr.

About This Item

UNSPSC Code:
12352204
NACRES:
NA.84

3.460,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

detection method

colorimetric

relevant disease(s)

hematological disorder; cancer

shipped in

wet ice

storage temp.

−20°C

General description

Isocitrate dehydrogenase (IDH) is an enzyme which catalyzes the interconversion of isocitrate and α-ketoglutarate. There are three IDH isoforms: IDH3 uses the cofactor NAD+ and catalyzes the third step in the citric acid cycle, while IDH1 and IDH2 use the cofactor NADP+ and catalyze the same reaction outside the citric acid cycle. This kit measures the activity of the IDH1 and IDH2 isoforms. Mutations in IDH1 and IDH2 have been linked with various brain tumors and acute myeloid leukemia.

Features and Benefits

  • Linear detection range 0.1 to 100 U/L for 30 minute reaction.
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for IDH activity determination in biological samples (e.g. plasma, serum, tissue and culture media.)

Principle

The Isocitrate Dehydrogenase (IDH) Assay Kit is a non-radioactive, colorimetric IDH assay based on the reduction of the tetrazolium salt MTT in a NADPH-coupled enzymatic reaction to a reduced form of MTT, which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is directly proportional to the enzyme activity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service