Skip to Content
Merck
All Photos(1)

Key Documents

M6513

Sigma-Aldrich

β-Melanocyte Stimulating Hormone human

≥95% (HPLC)

Synonym(s):

β-MSH human, β-Melanotropin human

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C118H174N34O35S
CAS Number:
Molecular Weight:
2644.92
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
form:
powder
Assay:
≥95% (HPLC)

Assay

≥95% (HPLC)

form

powder

solubility

water: 1 mg/mL, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H]2CCCN2C(=O)CNC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)N)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc3c[nH]cn3)C(=O)N[C@@H](Cc4ccccc4)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc5c[nH]c6ccccc56)C(=O)NCC(=O)N[C@@H](CO)C(=O)N7CCC[C@H]7C(=O)N8CCC[C@H]8C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(O)=O)C(O)=O

InChI

1S/C118H174N34O35S/c1-63(122)97(167)135-77(36-39-93(159)160)105(175)137-71(23-8-11-42-119)100(170)136-72(24-9-12-43-120)102(172)147-84(55-95(163)164)111(181)140-76(35-38-92(157)158)98(168)132-60-91(156)150-47-16-28-87(150)112(182)148-81(52-65-31-33-68(154)34-32-65)109(179)138-74(26-14-45-128-117(123)124)101(171)142-79(41-50-188-2)107(177)141-78(37-40-94(161)162)106(176)146-83(54-67-58-127-62-133-67)110(180)144-80(51-64-19-4-3-5-20-64)108(178)139-75(27-15-46-129-118(125)126)103(173)145-82(53-66-57-130-70-22-7-6-21-69(66)70)99(169)131-59-90(155)134-86(61-153)114(184)152-49-18-30-89(152)115(185)151-48-17-29-88(151)113(183)143-73(25-10-13-44-121)104(174)149-85(116(186)187)56-96(165)166/h3-7,19-22,31-34,57-58,62-63,71-89,130,153-154H,8-18,23-30,35-56,59-61,119-122H2,1-2H3,(H,127,133)(H,131,169)(H,132,168)(H,134,155)(H,135,167)(H,136,170)(H,137,175)(H,138,179)(H,139,178)(H,140,181)(H,141,177)(H,142,171)(H,143,183)(H,144,180)(H,145,173)(H,146,176)(H,147,172)(H,148,182)(H,149,174)(H,157,158)(H,159,160)(H,161,162)(H,163,164)(H,165,166)(H,186,187)(H4,123,124,128)(H4,125,126,129)/t63-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-/m0/s1

InChI key

XLLJFSCSXTYVTN-UPWOUFKKSA-N

Gene Information

human ... POMC(5443)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Glu-Lys-Lys-Asp-Glu-Gly-Pro-Tyr-Arg-Met-Glu-His-Phe-Arg-Trp-Gly-Ser-Pro-Pro-Lys-Asp

Biochem/physiol Actions

Hormone that stimulates melanogenesis; facilitates learning and memory; affects inflammatory and immune responses and peripheral nerve regeneration

Linkage

Derived from β-lipotropin (37-58)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service