Skip to Content
Merck
All Photos(1)

Key Documents

M6053

Sigma-Aldrich

Anti-Matrix Metalloproteinase-3, Propeptide Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MMP-3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 59 kDa
antigen (pro-form) 57 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP3(4314)

General description

Matrix metalloproteinase-3 (MMP-3) is also known as stromelysin 1 and the gene encoding it is localized on human chromosome 11q22.3.

Specificity

Reacts with reduced and to a lesser extent non-reduced MMP-3. The propeptide is shed after activation; therefore, the antibody recognizes the pro-form of MMP-3 and not the active forms. May be used to distinguish the latent from the active form of MMP-3. Recognizes Stromelysin-1 and Stromelysin-2, but not Stromelysin-3 or other MMP family members.

Immunogen

synthetic peptide corresponding to the propeptide region of human matrix metalloproteinase-3 (stromelysin-1).

Biochem/physiol Actions

Matrix metalloproteinase-3 (MMP-3) plays an important role in cartilage degradation. It can act on a large number of substrates like type II, III, IV, IX and X collagens, laminin and fibronectin. MMP-3 can also activate other matrix metalloproteinases like MMP-1,-2 and -13. It has been associated with rheumatoid arthritis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhitao Feng et al.
BMC musculoskeletal disorders, 15, 376-376 (2014-11-19)
Epidemiological studies have investigated the association between matrix metalloproteinase-3(MMP-3) gene-1171 5A/6A polymorphism and rheumatoid arthritis (RA), but the results were inconsistent. To evaluate the specific relationship, we performed a meta-analysis to clarify the controversies. The relevant literatures dated to December

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service