M4412
1H-1,2,4-Triazole-3-thiol
Synonym(s):
3-Mercapto-1,2,4-triazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C2H3N3S
CAS Number:
Molecular Weight:
101.13
Beilstein:
107731
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
mp
221-224 °C (lit.)
SMILES string
Sc1nc[nH]n1
InChI
1S/C2H3N3S/c6-2-3-1-4-5-2/h1H,(H2,3,4,5,6)
InChI key
AFBBKYQYNPNMAT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Quality
Photographic Grade
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A histochemical study of muscles of mice infected with Trichinella spiralis.
H N Awadalla et al.
Journal of the Egyptian Society of Parasitology, 12(1), 33-39 (1982-06-01)
Effect of some new antibilharzial drugs on oxygen consumption of mouse diaphragm and liver homogenates in experimental bilharziasis.
S M Amin et al.
Journal of the Egyptian Society of Parasitology, 12(1), 107-114 (1982-06-01)
Umut Salgin-Gökşen et al.
Bioorganic & medicinal chemistry, 15(17), 5738-5751 (2007-06-26)
Acetic acid hydrazide containing 5-methyl-2-benzoxazolinone (4) was synthesized by the condensation of 2-(5-methyl-2-benzoxazolinone-3-yl)acetate with hydrazine hydrate. Thiosemicarbazide derivatives (5a-5d) were afforded by the reaction of corresponding compound 4 with substituted isothiocyanates. The cyclization of compounds 5a-5d in the presence of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service