Skip to Content
Merck
All Photos(1)

Documents

L6520

Sigma-Aldrich

Prolactin from sheep pituitary

20-50 IU/mg

Synonym(s):

LTH, Lactogenic hormone from ovine pituitary glands, Luteotropic Hormone, PRL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

form

powder

specific activity

20-50 IU/mg

solubility

water: 7 mg/mL, clear to slightly hazy, colorless to faintly yellow

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

sheep ... PRL(443317)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Prolactin is a neuroendocrine hormone. The prolactin receptor is a transmembrane glycoprotein that belongs to the cytokine hematopoietic receptor family. A large number of cells and organs express the receptor, including B cells, T cells, macrophages, monocytes and neutrophils. Prolactin signal transduction involves the JAK/STAT families and the src kinase family. Induces lactation; inhibits secretion of gonadotropins; release is inhibited by dopamine.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharon R Ladyman et al.
eLife, 10 (2021-09-17)
As part of the maternal adaptations to pregnancy, mice show a rapid, profound reduction in voluntary running wheel activity (RWA) as soon as pregnancy is achieved. Here, we evaluate the hypothesis that prolactin, one of the first hormones to change
Jakub Sumbal et al.
Frontiers in cell and developmental biology, 8, 68-68 (2020-04-09)
Mammary gland development occurs mainly after birth and is composed of three successive stages: puberty, pregnancy and lactation, and involution. These developmental stages are associated with major tissue remodeling, including extensive changes in mammary epithelium, as well as surrounding stroma.
Gerda de Kuijper et al.
Research in developmental disabilities, 34(9), 2799-2809 (2013-06-25)
Individuals with intellectual disability frequently use antipsychotics for many years. This may have detrimental health effects, including neurological symptoms and metabolic and hormonal dysregulation, the latter possibly affecting bone metabolism. There is large variability in the degree in which antipsychotic
Xinyan Cao et al.
International journal of molecular sciences, 20(9) (2019-05-01)
Embryo implantation in the mink follows the pattern of many carnivores, in that preimplantation embryo diapause occurs in every gestation. Details of the gene expression and regulatory networks that terminate embryo diapause remain poorly understood. Illumina RNA-Seq was used to
Bibiana Moreno-Carranza et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 305(7), R720-R726 (2013-08-21)
Prolactin (PRL) is a potent liver mitogen and proangiogenic hormone. Here, we used hyperprolactinemic rats and PRL receptor-null mice (PRLR(-/-)) to study the effect of PRL on liver growth and angiogenesis before and after partial hepatectomy (PH). Liver-to-body weight ratio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service