Skip to Content
Merck
All Photos(1)

Documents

I7648

Nunc® inoculating loops

size 1 μL, pkg of 600 (50 sleeves of 12 each)

Synonym(s):

inoculating loop, inoculation loop, inoculation loops

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161502
NACRES:
NB.22

material

polystyrene

sterility

sterile

packaging

pkg of 600 (50 sleeves of 12 each)

manufacturer/tradename

Nunc 254410

size

1 μL

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

  • Accurate to semiquantitative standard for handling samples
  • Surface treated to enhance adhesion of the droplets
  • Reduced risk of spreading microorganisms since the loops are disposable
  • Non toxic

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service