Skip to Content
Merck
All Photos(1)

Documents

I1398

Sigma-Aldrich

Inhibin β-Subunit Fragment 67-94 human

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C150H240N36O43S2
CAS Number:
Molecular Weight:
3299.86
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

biological source

human

Assay

≥95% (HPLC)

form

powder

packaging

pkg of 0.5 mg

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](N)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@H]2CSSC[C@H](NC(=O)[C@@H](NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCCN)NC(=O)[C@@H]3CCCN3C(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@H](CCCCN)NC2=O)[C@@H](C)CC)C(C)C)C(C)C)[C@@H](C)O)C(=O)N[C@@H](CO)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc5c[nH]c6ccccc56)C(=O)NCC(=O)N[C@@H]([C@@H](C)CC)C(O)=O

InChI

1S/C150H240N36O43S2/c1-14-81(10)117(158)143(221)174-101(67-85-37-18-17-19-38-85)135(213)165-93(43-24-31-61-153)126(204)161-92(42-23-30-60-152)127(205)167-98(53-56-112(192)193)133(211)173-104(70-115(198)199)136(214)178-108-76-230-231-77-109(141(219)176-107(75-188)139(217)181-118(78(4)5)145(223)177-106(74-187)138(216)168-99(54-57-113(194)195)132(210)172-103(69-87-72-159-90-40-21-20-39-89(87)90)124(202)160-73-111(191)180-122(150(228)229)83(12)16-3)179-148(226)123(84(13)189)185-134(212)97(47-28-35-65-157)164-128(206)96(46-27-34-64-156)169-142(220)110-48-36-66-186(110)149(227)105(71-116(200)201)175-130(208)95(45-26-33-63-155)162-125(203)91(41-22-29-59-151)163-131(209)100(55-58-114(196)197)170-144(222)119(79(6)7)182-146(224)120(80(8)9)183-147(225)121(82(11)15-2)184-137(215)102(68-86-49-51-88(190)52-50-86)171-129(207)94(166-140(108)218)44-25-32-62-154/h17-21,37-40,49-52,72,78-84,91-110,117-123,159,187-190H,14-16,22-36,41-48,53-71,73-77,151-158H2,1-13H3,(H,160,202)(H,161,204)(H,162,203)(H,163,209)(H,164,206)(H,165,213)(H,166,218)(H,167,205)(H,168,216)(H,169,220)(H,170,222)(H,171,207)(H,172,210)(H,173,211)(H,174,221)(H,175,208)(H,176,219)(H,177,223)(H,178,214)(H,179,226)(H,180,191)(H,181,217)(H,182,224)(H,183,225)(H,184,215)(H,185,212)(H,192,193)(H,194,195)(H,196,197)(H,198,199)(H,200,201)(H,228,229)/t81-,82-,83-,84+,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,117-,118-,119-,120-,121-,122-,123-/m0/s1

InChI key

MFCCPILCNRYHNN-BLKMXGSCSA-N

Gene Information

human ... MSMB(4477)

Amino Acid Sequence

Ile-Phe-Lys-Lys-Glu-Asp-Cys-Lys-Tyr-Ile-Val-Val-Glu-Lys-Lys-Asp-Pro-Lys-Lys-Thr-Cys-Ser-Val-Ser-Glu-Trp-Gly-Ile

General description

Inhibin β-Subunit (also called as MSMB, microseminoprotein-β) is a cysteine-rich and nonglycosylated protein. It is one of the major components of semen.

Biochem/physiol Actions

Inhibin β-Subunit (also called as MSMB, microseminoprotein-β) is upregulated in ovarian cancer and is correlated with the levels of prostasin. Low expression of MSMB is linked with increased risk of prostate cancer. It is involved in the inhibition of FSH (follicle stimulating hormone), thereby abolishing growth-enhancing activity of FSH in prostate cancer.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marni Stott-Miller et al.
The Prostate, 73(16), 1803-1809 (2013-09-17)
Recently, a genetic variant (rs10993994) in the MSMB gene associated with prostate cancer (PCa) risk was shown to correlate with reduced prostate secretory protein of 94 amino acids (PSP94) levels. Although the biological activity of PSP94 is unclear, one of
Nicholas Shukeir et al.
Cancer research, 63(9), 2072-2078 (2003-05-03)
Prostate cancer is a common malignancy affecting men, which is often associated with skeletal metastases resulting in significant morbidity and mortality. In this hormone-dependent cancer, low levels of a prostate secretory protein of 94 amino acids (PSP-94) are associated with
N J Arbatti et al.
FEBS letters, 181(1), 57-63 (1985-02-11)
The complete synthesis of the C-terminal 28 residues segment 67-94 of human seminal plasma beta-Inhibin, called beta 2-Inhibin, is reported. The Inhibin-like activity of the native 94 amino acids beta-Inhibin is compared to that of the synthetic replica of beta
S D Mahale et al.
International journal of peptide and protein research, 42(2), 132-137 (1993-08-01)
Observation of contradictory results with the in vitro assays for inhibin-like activity of the carboxyl terminal 28 amino acid peptide 67-94 with a disulfide loop, of human seminal plasma inhibin (HSPI), prompted us to synthesize both the linear and the
J-x Ma et al.
Cell death & disease, 5, e1407-e1407 (2014-09-05)
Ovarian cancer is a leading cause of cancer death as diagnosis is frequently delayed to an advanced stage. Effective biomarkers and screening strategies for early detection are urgently needed. In the current study, we identify PSP94 as a key upstream

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service