HS120249
Wide mouth wash bottle
capacity 500 mL, blue PE-LD bottle
Select a Size
283,00 kr.
Select a Size
About This Item
283,00 kr.
Recommended Products
material
blue PE-LD bottle
polypropylene cap
feature
label no label
vented
with cap
packaging
pack of 6 ea
manufacturer/tradename
Heathrow Scientific HS120249
capacity
500 mL
Related Categories
General description
The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
- Wide bottle mouth makes filling easier
- Color-coding for user, application or location identification
- Bottle is made from LDPE, with a PP closure
- 500 mL bottle size,
- Supplied in packs of 6
Choose from one of the most recent versions:
Certificates of Analysis (COA)