Skip to Content
Merck
All Photos(1)

Documents

HPA021872

Sigma-Aldrich

Anti-PTPRE antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein-tyrosine phosphatase epsilon, Anti-R-PTP-epsilon, Anti-Receptor-type tyrosine-protein phosphatase epsilon

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

VMLTEVQEREQDKCYQYWPTEGSVTHGEITIEIKNDTLSEAISIRDFLVTLNQPQARQEEQVRVVRQFHFHGWPEIGIPAEGKGMIDLIAAVQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPRE(5791)

General description

PTPRE (Protein tyrosine phosphatase, receptor type, E) is a transmembrane protein consisting of a short extracellular domain and two tandemly repeated intracellular PTPase domains. It is located on chromosome 10q26 in humans. It is expressed in testis, brain, lung and lymph node. Its expression has also been observed during neuronal differentiation and in murine mammary tumour cells. It has an isoform consisting of an N-terminal catalytic, active phosphatase domain 1 (PD1).

Immunogen

Receptor-type tyrosine-protein phosphatase epsilon Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PTPRE (Protein tyrosine phosphatase, receptor type, E) is associated with various signalling pathways including down regulation of insulin receptor signalling and Janus kinase (Jak)-STAT inhibition signalling pathway. It also plays an important role in the regulation of ERK1/2 pathway by emerging as a phosphatase through its catalytic activity. Study shows that it may play an accessory role in tumour development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72873

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Wabakken et al.
Scandinavian journal of immunology, 56(3), 276-285 (2002-08-24)
This report describes the identification and characterization of a novel cytoplasmic isoform of human protein tyrosine phosphatase epsilon (PTPepsilon). The novel isoform, denoted cyt-PTPepsilonPD1, displays only the N-terminal catalytic, active phosphatase domain 1 (PD1) which is common in all known
T Wabakken et al.
Scandinavian journal of immunology, 56(2), 195-203 (2002-07-18)
The expression of protein tyrosine phosphatase epsilon (PTPepsilon) was studied in human tissues and blood cells. High mRNA expression was observed in peripheral blood leucocytes, particularly in monocytes and granulocytes which revealed at least four distinct transcripts. In lymphocytes, PTPepsilon
Assignment of the human protein tyrosine phosphatase epsilon (PTPRE) gene to chromosome 10q26 by fluorescence in situ hybridization.
A M van den Maagdenberg et al.
Genomics, 30(1), 128-129 (1995-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service