Skip to Content
Merck
All Photos(2)

Key Documents

HPA020444

Sigma-Aldrich

Anti-PAK5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KIAA1264, Anti-PAK7, Anti-PAK-5, Anti-PAK-7, Anti-Serine/threonine-protein kinase PAK 7, Anti-p21-activated kinase 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

KPSEYSDLKWEYQRASSSSPLDYSFQFTPSRTAGTSGCSKESLAYSESEWGPSLDDYDRRPKSSYLNQTSPQPTMRQRSRSGSGLQEPMMPFGASAFKTHPQGHSYNSYTYPRLSEPTMCIPKVDYDRAQMVLSP

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAK7(57144)

General description

p21 protein (Cdc42/Rac)-activated kinase 7 (PAK7) is part of the Rac (ras-related protein)/Cdc42 (cell division cycle 42)-associated serine/threonine protein kinase family. It is expressed in the brain and contains regulatory and kinase domains. The gene encoding PAK7 is localized on human chromosome 20p12.

Immunogen

p21 (RAC1) activated kinase 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

p21 protein (Cdc42/Rac)-activated kinase 7 (PAK7) interacts with GTPases like cell division control protein 42 (Cdc42) and stimulates mitogen-activated protein kinase (MAPK) signaling pathways. PAK7 also phosphorylates Raf1 (serine/threonine-protein kinase) and thus aids its translocation to the mitochondria. Overexpression of this protein is seen in colorectal and pancreatic cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70075

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun Han et al.
Cell biology international, 38(12), 1394-1402 (2014-07-24)
p21-activated kinase 7 (PAK7), also named as PAK5, is a member of Rac/Cdc42-associated Ser/Thr protein kinases. It is overexpressed in some types of cancer such as colorectal and pancreatic cancers. However, the expression status and biological function of PAK7 in
Sophie Cotteret et al.
Molecular and cellular biology, 26(8), 3215-3230 (2006-04-04)
p21-activated kinase 5 (Pak5) is an effector for the small GTPase Cdc42, known to activate cell survival signaling pathways. Previously, we have shown that Pak5 localizes primarily to mitochondria. To study the relationship between Pak5 localization and its effects on
Xuefeng Gu et al.
International journal of biological sciences, 11(2), 230-237 (2015-01-30)
Glioma is the most common type of primary intracranial tumor and is highly lethal due to its pathogenetic location, high invasiveness, and poor prognosis. Even combined surgery and chemoradiotherapy do not effectively rescue glioma patients. Molecular target therapy is considered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service