Skip to Content
Merck
All Photos(7)

Key Documents

HPA018166

Sigma-Aldrich

Anti-RRP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Novel nuclear protein 1, Anti-Nucleolar protein Nop52, Anti-Protein NNP-1, Anti-RRP1-like protein, Anti-Ribosomal RNA processing protein 1 homolog A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
4.340,00 kr.

4.340,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
4.340,00 kr.

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

4.340,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

NLKFIDPFCRIAARTKDSLVLNNITRGIFETIVEQAPLAIEDLLNELDTQDEEVASDSDESSEGGERGDALSQKRSEKPPA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RRP1(8568)

General description

The gene RRP1 (ribosomal RNA processing protein-1 homolog A) has been mapped to human chromosome 21q22.3. The protein is a human homologue of the yeast ribosomal RNA processing protein RRP1. The protein has been localized to the nucleoli and at the periphery of the chromosomes.

Immunogen

Ribosomal RNA processing protein 1 homolog A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In nucleoli, ribosomal RNA processing protein-1 homolog A (RRP1) is excluded from the rRNA transcription sites, which accumulates in the granular external domain and mainly co-localizes with nucleolar proteins such as B23 which are involved in late processing step. The perinuclear bodies sequentially recruit fibrillarin, nucleoli and RRP1 together with B23 during formation of the nucleolus at the end of mitosis. p32, fibrillarin and RRP1 play key roles in the separation of pre-90S particle into pre-40S and pre-60S particles. RRP1 is also involved in the cleavage at site 2 in internal transcribed space1 of pre-rRNAs at early stages of human ribosome biogenesis. Together the above two events are essential to produce the small and large ribosomal subunits in human cells.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73884

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T M Savino et al.
The Journal of cell biology, 153(5), 1097-1110 (2001-05-31)
To understand how nuclear machineries are targeted to accurate locations during nuclear assembly, we investigated the pathway of the ribosomal RNA (rRNA) processing machinery towards ribosomal genes (nucleolar organizer regions [NORs]) at exit of mitosis. To follow in living cells
Edward W Horsey et al.
RNA (New York, N.Y.), 10(5), 813-827 (2004-04-22)
The Saccharomyces cerevisiae gene RRP1 encodes an essential, evolutionarily conserved protein necessary for biogenesis of 60S ribosomal subunits. Processing of 27S pre-ribosomal RNA to mature 25S rRNA is blocked and 60S subunits are deficient in the temperature-sensitive rrp1-1 mutant. We
Uta Behrends et al.
International journal of cancer, 100(6), 669-677 (2002-09-05)
Autologous serological screening of a cDNA expression library (SEREX) derived from childhood neuroblastoma led to the identification of 10 different antigens, including 6 novel gene products. The novel antigen 018INX was derived from a small open reading frame in a
Harunori Yoshikawa et al.
Molecular & cellular proteomics : MCP, 10(8), M110-M110 (2011-05-04)
Ribosome biogenesis starts with transcription of the large ribosomal RNA precursor (47S pre-rRNA), which soon combines with numerous factors to form the 90S pre-ribosome in the nucleolus. Although the subsequent separation of the pre-90S particle into pre-40S and pre-60S particles
T M Savino et al.
Journal of cell science, 112 ( Pt 12), 1889-1900 (1999-05-26)
We report the molecular characterization of a novel nucleolar protein, Nop52, and its subcellular distribution during the cell cycle and nucleologenesis. This protein was originally identified with human autoantibodies which were subsequently used to clone its corresponding cDNA. Transfection experiments

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service