Skip to Content
Merck
All Photos(3)

Key Documents

HPA018157

Sigma-Aldrich

Anti-PAFAH2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HSD-PLA2, Anti-Platelet-activating factor acetylhydrolase 2, cytoplasmic, Anti-Serine-dependent phospholipase A2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
4.340,00 kr.

4.340,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
4.340,00 kr.

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

4.340,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

RGFVVAVPEHRDRSAATTYFCKQAPEENQPTNESLQEEWIPFRRVEEGEKEFHVRNPQVHQRVSECLRVLKILQEVTAGQTVFNILPGGLDLMTLKGNIDMSRVAVMGHS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAFAH2(5051)

General description

Platelet-activating factor acetylhydrolases (PAFAHs) are a family of specialized phospholipase A2 enzymes. PAFAH2 is a 40kDa monomer with most abundant expression in bovine liver and kidney. In resting conditions the protein is localized in the cytoplasm as homodimers, and it responds to oxidative stress by binding to endoplasmic reticulum and Golgi membranes as monomers.

Immunogen

Platelet-activating factor acetylhydrolase 2, cytoplasmic recombinant protein epitope signature tag (PrEST)

Application

Anti-PAFAH2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Platelet-activating factor acetylhydrolases2 (PAFAH2) hydrolyzes platelet-activating factor and oxidatively fragmented phospholipids at the sn-2 position, protecting cells from oxidative stress induced apoptosis. PAFAH2 is up-regulated in patients with extensive coronary artery disease. Overexpression of PAFAH2 protects human keratinocyte-derived cells against apoptosis induced by t-butylhydroperoxide (t-BuOOH) and ultraviolet B radiation. Similarly, overexpression of PAFAH2 in Chinese hamster ovary cells inhibits apoptosis induced by the pro-oxidant t-BuOOH. Presence of PAFAH2 in mice models exerts strong neuroprotective effects against ischemic injury in the central nervous system.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76147

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth S Monillas et al.
Biochimica et biophysica acta, 1854(5), 469-475 (2015-02-25)
The intracellular enzyme platelet-activating factor acetylhydrolase type-II (PAFAH-II) hydrolyzes platelet-activating factor and oxidatively fragmented phospholipids. PAFAH-II in its resting state is mainly cytoplasmic, and it responds to oxidative stress by becoming increasingly bound to endoplasmic reticulum and Golgi membranes. Numerous
XiaoFei Li et al.
Atherosclerosis, 218(1), 83-89 (2011-05-25)
Inflammation plays a crucial role in plaque vulnerability. Calcifications can be detected by means of in vivo imaging techniques. The study purpose is to assess a potential association between tissue localization of calcifications and the inflammatory biomarkers, C-reactive protein (CRP)
Kimiko Umemura et al.
Stroke, 38(3), 1063-1068 (2007-02-03)
Platelet-activating factor (PAF) and oxidized unsaturated free fatty acids have been postulated to aggravate neuronal damage in the postischemic brain. Type II PAF-acetylhydrolase (PAF-AH II) not only terminates signals by PAF by its PAF-hydrolyzing activity but also protects cells against
Erik Berg Schmidt et al.
Atherosclerosis, 196(1), 420-424 (2006-12-13)
Lipoprotein-associated phospholipase A(2) (Lp-PLA(2)) is an emerging risk factor for cardiovascular disease. In the present study, plasma levels of Lp-PLA(2) were measured in patients (n=301) admitted to elective coronary angiography because of suspected coronary artery disease (CAD). In a multiple
A Matsuzawa et al.
The Journal of biological chemistry, 272(51), 32315-32320 (1998-01-24)
Platelet-activating factor-acetylhydrolase (PAF-AH), which removes the acetyl group at the sn-2 position of PAF, is distributed widely in tissues and plasma. Tissue cytosol contains at least two types of PAF-AH, isoforms Ib and II. Isoform Ib is a tertiary G-protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service