Skip to Content
Merck
All Photos(1)

Key Documents

HPA015638

Sigma-Aldrich

Anti-ADGRE3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EMR3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

NASCVNNTHCTCNHGYTSGSGQKLFTFPLETCNDINECTPPYSVYCGFNAVCYNVEGSFYCQCVPGYRLHSGNEQFSNSNENTCQDTTSSKTTEGRKELQKIVDKFESLLTNQTLWRTEGRQEISSTATTILRDVESKVL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EMR3(84658)

Immunogen

adhesion G protein-coupled receptor E3

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

EGF-like module-containing mucin-like hormone receptor-like 3 is a protein encoded by the ADGRE3 gene in humans and is mapped to chromosome 19p13.1. It belongs to epidermal growth factor-seven-transmembrane (EGF-TM7) family of adhesion class TM7 receptors. It is expressed on granulocytes. It is a G-protein coupled receptor and may have cellular function. Overexpression of this gene is associated with glioblastoma (GBM) multiforme. This protein also plays a role in myeloid-myeloid interactions during immune and inflammatory responses.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72615

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Stacey et al.
The Journal of biological chemistry, 276(22), 18863-18870 (2001-03-30)
The epidermal growth factor (EGF)-TM7 subgroup of G-protein-coupled receptors is composed predominantly of leukocyte-restricted glycoproteins defined by their unique hybrid structure, in which extracellular EGF-like domains are coupled to a seven-span transmembrane moiety via a mucin-like stalk. The EGF-TM7 group
Ari J Kane et al.
Neuroreport, 21(16), 1018-1022 (2010-09-10)
Epidermal growth factor module-containing mucin-like hormone receptor-3 (EMR-3) is a G-protein coupled receptor with unknown ligand and cellular function. Upregulation of EMR-3 in glioblastoma (GBM) multiforme is associated with poor survival. We investigated the expression patterns and functional significance of
Mourad Matmati et al.
Journal of leukocyte biology, 81(2), 440-448 (2006-11-17)
EMR3 is a member of the epidermal growth factor-seven-transmembrane (EGF-TM7) family of adhesion class TM7 receptors. This family also comprises CD97, EMR1, EMR2, and EMR4. To characterize human EMR3 at the protein level, we generated Armenian hamster mAb. Using the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service