Skip to Content
Merck
All Photos(3)

Key Documents

HPA011862

Sigma-Aldrich

Anti-ENG antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD105 antigen, Anti-Endoglin precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
3.850,00 kr.

3.850,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
3.850,00 kr.

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

3.850,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ASFVELPLASIVSLHASSCGGRLQTSPAPIQTTPPKDTCSPELLMSLIQTKCADDAMTLVLKKELVAHLKCTITGLTFWDPSCEAEDRGDKFVLRSAYSSCGMQVSASMISNEAVVNILSSSSPQRK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ENG(2022)

Immunogen

Endoglin precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ENG antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ENG (Endoglin) is a membranous, 180kDa, disulfide-linked transmembrane protein. This homodimeric protein is expressed on proliferating endothelial cells. It is a member of the transforming growth factor β superfamily and essential for the formation as well as complete development of the blood cells. Its overexpression have been reported in the various tumor-associated endothelial cells, chemoresistant populations of ovarian cancer cells, and leukemia, gastrointestinal stromal tumors (GIST), melanoma, and laryngeal cancers. During extracellular matrix (ECM) expression, it functions as a TGF-β co-receptor and negatively control the ECM protein expressions. Studies show the association of ENG with the chronic kidney fibrosis disease. It also participates in the BMP9 signaling pathway in endothelial cells and modulates TGF-β1 signaling through SMAD3. Mutation analysis of the ENG shows the vascular defect known as hereditary hemorrhagic telangiectasia (HHT1).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71767

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoe Numakura et al.
Anticancer research, 39(1), 387-393 (2018-12-29)
Spindle-shaped stromal cells of tumors are derived from various cellular origins, including mesenchymal stem cells (MSCs). MSCs express CD73, CD90 and CD105 antigens. Herein, the aim was to investigate the association between the expression of specific MSC markers in gastric
Jose M Muñoz-Felix et al.
Expert reviews in molecular medicine, 16, e18-e18 (2014-12-03)
Tubulointerstitial fibrosis and glomerulosclerosis, are a major feature of end stage chronic kidney disease (CKD), characterised by an excessive accumulation of extracellular matrix (ECM) proteins. Transforming growth factor beta-1 (TGF-β1) is a cytokine with an important role in many steps
Roselyne Castonguay et al.
The Journal of biological chemistry, 286(34), 30034-30046 (2011-07-09)
Endoglin (CD105), a transmembrane protein of the transforming growth factor β superfamily, plays a crucial role in angiogenesis. Mutations in endoglin result in the vascular defect known as hereditary hemorrhagic telangiectasia (HHT1). The soluble form of endoglin was suggested to
Olivier Nolan-Stevaux et al.
PloS one, 7(12), e50920-e50920 (2013-01-10)
Endoglin (ENG), a co-receptor for several TGFβ-family cytokines, is expressed in dividing endothelial cells alongside ALK1, the ACVRL1 gene product. ENG and ACVRL1 are both required for angiogenesis and mutations in either gene are associated with Hereditary Hemorrhagic Telangectasia, a
Angela J Ziebarth et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 19(1), 170-182 (2012-11-14)
Endoglin (CD105) is a membranous protein overexpressed in tumor-associated endothelial cells, chemoresistant populations of ovarian cancer cells, and potentially stem cells. Our objective was to evaluate the effects and mechanisms of targeting endoglin in ovarian cancer. Global and membranous endoglin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service