Skip to Content
Merck
All Photos(5)

Key Documents

HPA010831

Sigma-Aldrich

Anti-PHF2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GRC5, Anti-PHD finger protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
4.340,00 kr.

4.340,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
4.340,00 kr.

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

4.340,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

LEIREQTKSKSEAKWKYKNSKPDSLLKMEEEQKLEKSPLAGNKDNKFSFSFSNKKLLGSKALRPPTSPGVFGALQNFKEDKPKPVRDEYEYVSDDGELKIDEFPIRRKKNAPKRDLSFLLDKKAVLPTPVTKPKLD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHF2(5253)

General description

PHF2 (PHD finger protein 2) is a member of α-ketoglutarate-Fe2+-dependent dioxygenases. This family of proteins contains three members namely, PHF2, PHF8 and KIAA1718. PHF2 gene maps to human chromosome 9q22. It contains a Jumonji domain and a plant homeodomain (PHD) at its N-terminal. Its C-terminal contains four repeats of TPAST sequence, along with a serine and threonine rich sequence. It has two possible PEST sequences and eight putative nuclear localization signals. It is widely expressed in most adult tissues.

Immunogen

PHD finger protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PHF2 (PHD finger protein 2) is a histone H3K9 demethylase that demethylates H3K9me1, and through its PHD (plant homeodomain) domain interacts with H3K4me3. Therefore, it regulates the demethylation of histone. However, it remains inactive unless phosphorylated by protein kinase A (PKA). Once activated, it forms a complex with a DNA-binding protein called ARID5B (AT rich interactive domain 5B). This causes demethylation of ARID5B, and PHF2- ARID5B complex then interacts with the target promoters. Hence, this complex regulates histone methylation and gene transcription. In mouse embryos, PHF2 is expressed in higher levels in neural tube and dorsal root ganglia, as opposed to its expression in other tissues. This expression profile suggests that PHF2 might have a role in hereditary sensory neuropathy type I (HSN1). This protein is found to be associated with many cancers, and its expression is up-regulated in esophageal squamous cell carcinoma (ESCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71317

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling-Ling Sun et al.
Acta histochemica, 115(1), 56-62 (2012-04-27)
Jumonji AT-rich interactive domain 1B (JARID1B) and PHD finger protein 2 (PHF2), members of the histone demethylases, have been found to be involved in many types of tumors. However, the expression and prognostic significance of JARID1B and PHF2 in esophageal
Lu Zhang et al.
RSC advances, 8(69), 39520-39528 (2018-11-27)
PHD Finger Protein 2 (PHF2), as a protein code and a transcription regulatory gene, is a member of the Jumonji-C domain (JmjC). PHF2 is located at human chromosome 9q22.31 and is frequently decreased in various malignancies. However, the definite role
Ying Dong et al.
Signal transduction and targeted therapy, 8(1), 95-95 (2023-03-06)
Epithelial to mesenchymal transition (EMT) plays a crucial role in cancer metastasis, accompanied with vast epigenetic changes. AMP-activated protein kinase (AMPK), a cellular energy sensor, plays regulatory roles in multiple biological processes. Although a few studies have shed light on
K Hasenpusch-Theil et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 10(3), 294-298 (1999-03-02)
We have isolated and characterized a novel PHD finger gene, PHF2, which maps to human Chromosome (Chr) 9q22 close to D9S196. Its mouse homolog was also characterized and mapped to the syntenic region on mouse Chr 13. The predicted human
Atsushi Baba et al.
Nature cell biology, 13(6), 668-675 (2011-05-03)
Reversible histone methylation and demethylation are highly regulated processes that are crucial for chromatin reorganization and regulation of gene transcription in response to extracellular conditions. However, the mechanisms that regulate histone-modifying enzymes are largely unknown. Here, we characterized a protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service