Skip to Content
Merck
All Photos(5)

Documents

HPA008706

Sigma-Aldrich

Anti-DPP8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DP8 antibody produced in rabbit, Anti-DPRP-1 antibody produced in rabbit, Anti-Dipeptidyl peptidase 8 antibody produced in rabbit, Anti-Dipeptidyl peptidase IV-related protein 1 antibody produced in rabbit, Anti-Dipeptidyl peptidase VIII antibody produced in rabbit, Anti-Prolyl dipeptidase DPP8 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MAAAMETEQLGVEIFETADCEENIESQDRPKLEPFYVERYSWSQLKKLLADTRKYHGYMMAKAPHDFMFVKRNDPDGPHSDRIYYLAMSGENRENTLFYSEIPKTINRAAVLMLSWKPL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DPP8(54878)

General description

DPP8 (dipeptidyl-peptidase 8) is a dipeptidyl aminopeptidase, which cleaves in a post-proline manner, and is a homolog of DPPIV and FAP (fibroblast activation protein). This gene is localized to human chromosome 15q22, and codes for a protein of 882 amino acids. This gene has a ubiquitous expression pattern. It is localized to the cytoplasm and has a molecular weight of 100kDa. It lacks N- and O-glycosylation sites and transmembrane region. It belongs to the DPPIV gene family.

Immunogen

Dipeptidyl peptidase 8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DPP8 (dipeptidyl-peptidase 8) is thought to have immunological roles, with involvement in lung allergic responses and bowel inflammatory disorders. It is up-regulated in human meningiomas, and its mRNa is found in chronic lymphocytic leukemia. It acts as a survival factor in Ewing sarcoma family of tumors. It also cleaves neuropeptide Y, glucagon-like peptide-2 and peptide YY. It is involved in the pathogenesis of liver disease, where it is implicated in lymphocyte activation and apoptosis. In atherosclerosis, this protein has a high expression in macrophage-dominant regions of plaque. As macrophages play a prominent role in atherosclerosis pathogenesis, this protein might be involved in the same.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71650

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumaiya Chowdhury et al.
World journal of gastroenterology, 19(19), 2883-2893 (2013-05-25)
To investigate the expression of dipeptidyl peptidase (DPP) 8 and DPP9 in lymphocytes and various models of liver fibrosis. DPP8 and DPP9 expression were measured in mouse splenic CD4⁺ T-cells, CD8⁺ T-cells and B-cells (B220⁺), human lymphoma cell lines and
Veerle Matheeussen et al.
Basic research in cardiology, 108(3), 350-350 (2013-04-24)
Atherosclerosis is a chronic inflammatory disorder of the arterial wall leading to coronary artery disease, stroke, and peripheral arterial disease. Along with the discovery of dipeptidyl peptidase 4 (DPP4) as a therapeutic target in type 2 diabetes, a role for
C A Abbott et al.
European journal of biochemistry, 267(20), 6140-6150 (2000-09-30)
Dipeptidyl peptidase (DPP) IV has roles in T-cell costimulation, chemokine biology, type-II diabetes and tumor biology. Fibroblast activation protein (FAP) has been implicated in tumor growth and cirrhosis. Here we describe DPP8, a novel human postproline dipeptidyl aminopeptidase that is
Claire H Wilson et al.
The Journal of biological chemistry, 288(20), 13936-13949 (2013-03-23)
Dipeptidyl peptidases (DP) 8 and 9 are homologous, cytoplasmic N-terminal post-proline-cleaving enzymes that are anti-targets for the development of DP4 (DPPIV/CD26) inhibitors for treating type II diabetes. To date, DP8 and DP9 have been implicated in immune responses and cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service