Skip to Content
Merck
All Photos(2)

Key Documents

HPA007326

Sigma-Aldrich

Anti-GPR61 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Biogenic amine receptor-like G-protein coupled receptor, Anti-Probable G-protein coupled receptor 61

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
4.340,00 kr.

4.340,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
4.340,00 kr.

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

4.340,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SMFRVARVAAMQHGPLPTWMETPRQRSESLSSRSTMVTSSGAPQTTPHRTFGGGKAAVV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR61(83873)

Immunogen

Probable G-protein coupled receptor 61 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GPR61 (G-protein coupled receptor 61) gene is a member of the G-protein coupled receptor 1 family. The gene encodes an orphan receptor that is predominantly expressed in the brain, implicating its role in several physiological functions in the central nervous system. GPR61 deficiency in mice causes obesity associated with hyperphagia, which suggests that it may be involved in the regulation of food-intake and body weight. It may serve as a therapeutic target during the treatment of obesity or eating disorders. The N-terminal domain of this protein that functions as a tethered intramolecular ligand is found to be essential for its constitutive activity. The function of this gene in humans is yet to be elucidated.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71176

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaru Toyooka et al.
Journal of neuroscience research, 87(6), 1329-1333 (2008-11-26)
G-protein-coupled receptor 61 (GPR61) is an orphan receptor that is abundantly expressed in the brain, which suggests its involvement in various physiological functions in the central nervous system. It couples with Gs and shows constitutive activity. To investigate the role
Hirohide Nambu et al.
Life sciences, 89(21-22), 765-772 (2011-10-06)
GPR61 is an orphan G protein-coupled receptor whose function remains unknown. The purpose of the present study is to elucidate the importance of GPR61 in metabolism by characterization of GPR61-deficient mice. Male GPR61-deficient mice were characterized regarding various metabolic parameters

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service