H9392
Heparin disaccharide III-S sodium salt
Synonym(s):
α-ΔUA-2S-[1→4]-GlcNS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C12H16NO16S2Na3
CAS Number:
Molecular Weight:
563.35
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Recommended Products
biological source
Porcine
Assay
≥98%
form
powder
technique(s)
HPLC: suitable
storage temp.
−20°C
SMILES string
[Na].OCC(O)C(OC1OC(=CC(O)C1OS(O)(=O)=O)C(O)=O)C(O)C(NS(O)(=O)=O)C=O
InChI
1S/C12H19NO16S2.Na.H/c14-2-4(13-30(21,22)23)8(18)9(6(17)3-15)28-12-10(29-31(24,25)26)5(16)1-7(27-12)11(19)20;;/h1-2,4-6,8-10,12-13,15-18H,3H2,(H,19,20)(H,21,22,23)(H,24,25,26);;
InChI key
PYSLWUAQYWXSFK-UHFFFAOYSA-N
Biochem/physiol Actions
Products of the digestion of heparin and heparan sulfate by various heparinases
Other Notes
In the following listings, ΔUA = 4-deoxy-L-threo-hex-4-enopyranosyluronic acid; GlcN = D-glucosamine; Ac = Acetyl; NS, 2S, 6S, = N-sulfo, 2-sulfate and 6-sulfate respectively.
Preparation Note
Produced by the action of heparinase I and II
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service