Skip to Content
Merck
All Photos(1)

Key Documents

H4760

Sigma-Aldrich

17α-Hydroxyprogesterone 17-acetate 3-(O-carboxymethyl)oxime

Synonym(s):

17α-Hydroxy-4-pregnene-3,20-dione 3-(O-carboxymethyl)oxime 17-acetate, 17α-Hydroxyprogesterone 3-(O-carboxymethyl)oxime 17-acetate, 4-Pregnene-3,20-dione-17α-ol 3-(O-carboxymethyl)oxime 17-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H35NO6
CAS Number:
Molecular Weight:
445.55
UNSPSC Code:
12352200
PubChem Substance ID:

InChI

1S/C25H35NO6/c1-15(27)25(32-16(2)28)12-9-21-19-6-5-17-13-18(26-31-14-22(29)30)7-10-23(17,3)20(19)8-11-24(21,25)4/h13,19-21H,5-12,14H2,1-4H3,(H,29,30)/b26-18+/t19-,20+,21+,23+,24+,25+/m1/s1

InChI key

NIJDYRWVCFKWIL-IEMAJDPISA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service