Skip to Content
Merck
All Photos(1)

Key Documents

H156

Sigma-Aldrich

(+)-3-Hydroxymorphinan hydrobromide

neurotrophic to dopaminergic neurons

Synonym(s):

(+)-9α,13α,14α-Morphinan-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO · HBr
CAS Number:
Molecular Weight:
324.26
UNSPSC Code:
12352204
PubChem Substance ID:

description

neurotrophic to dopaminergic neurons

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

Br.Oc1ccc2C[C@@H]3NCC[C@]4(CCCC[C@H]34)c2c1

InChI

1S/C16H21NO.BrH/c18-12-5-4-11-9-15-13-3-1-2-6-16(13,7-8-17-15)14(11)10-12;/h4-5,10,13,15,17-18H,1-3,6-9H2;1H/t13-,15+,16+;/m1./s1

InChI key

LJAQABDSIONCIU-KHUAAREISA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Vetticaden et al.
Pharmaceutical research, 6(1), 13-19 (1989-01-01)
Polymorphic differences in dextromethorphan metabolism were observed in three studies conducted in a total of 44 subjects (of Dutch origin) administered 60 mg dextromethorphan hydrobromide as an OROS tablet. Mean plasma dextromethorphan (DM) concentrations after a single dose and at
Timothy S Tracy et al.
American journal of obstetrics and gynecology, 192(2), 633-639 (2005-02-08)
The purpose of this study was to determine whether drug metabolism (CYP1A2, CYP2D6 and CYP3A) activity varies in the pregnant state compared with the nonpregnant state. Subjects were studied at 14 to18 weeks of gestation, 24 to 28 weeks of
Eun-Joo Shin et al.
Journal of pharmacological sciences, 106(1), 22-27 (2008-01-17)
Drug abuse involving dextromethorphan, an antitussive, has been a social problem in various geographic locations since the 1960s. Ironically, high doses of the drug confer neuroprotective activity with central nervous system and behavioral effects. Accumulating evidence suggests that metabolism to
Tzu-Yu Lin et al.
Neurochemistry international, 54(8), 526-534 (2009-05-12)
Dextromethorphan (DM), a widely used antitussive, has demonstrated an effective neuroprotective effect. Excessive release of glutamate is considered to be an underlying cause of neuronal damage in several neurological diseases. In the present study, we investigated whether DM or its
Wei Zhang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 20(14), 2496-2511 (2006-12-05)
We investigated the neuroprotective property of analogs of dextromethorphan (DM) in lipopolysaccharide (LPS) and 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) models to identify neuroprotective drugs for Parkinson's disease (PD). In vivo studies showed that daily injections with DM analogs protected dopamine (DA) neurons in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service