GE38152103010350
Sera-Mag™ Magnetic Oligo(dt) particles
Cytiva 38152103010350, 1 μm avg. part. size, suspension (1% Solids)
Select a Size
24.640,00 kr.
Select a Size
About This Item
24.640,00 kr.
Recommended Products
ligand
(Olido(dt) particles)
form
suspension (1% Solids)
packaging
pkg of 100 mL
manufacturer/tradename
Cytiva 38152103010350
concentration
10 mg/mL
avg. part. size
1 μm
shipped in
wet ice
storage temp.
2-8°C
Application
Features and Benefits
- Carboxylic groups on the surface permit easy covalent coupling to biomolecules of interest using convenient carbodiimide chemistry.
- Different levels of hydrophobicity/hydrophilicity available.
- Available in Sera-Mag™ SpeedBeads and original Sera-Mag™ versions.
- Excellent sensitivity and low nonspecific binding for greater accuracy.
- Fast Magnetic Response Times and Stable Physical Integrity
Analysis Note
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service