GE17-5317-02
Capto™ MMC
Cytiva 17-5317-02, pack of 100 mL
Synonym(s):
Capto MMC Resin, MMC Chromatography Resin
About This Item
Recommended Products
description
Ion Exchanger Type (value)
packaging
pack of 100 mL
manufacturer/tradename
Cytiva 17-5317-02
matrix
highly cross-linked agarose
avg. part. size
75 μm (d50v)
cleaning in place
2-14
working range
2-12
capacity
>45 mg binding capacity (BSA/ml medium at 30 mS/cm)
suitability
suitable for bioprocess medium
Related Categories
General description
Capto™ MMC is a multimodal cation exchanger, belonging to the Capto™ family of BioProcess media for fast, efficient and cost-effective protein purification. Capto™ MMC combines agarose base matrix developments with innovative ligand chemistry. It is composed of a rigid, high-flow agarose matrix and a novel multimodal ligand. The multimodal functionality gives Capto™ MMC a different selectivity, compared with traditional ion exchangers. It can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges at both high and low ionic strengths.
The ligand contains caroxylic group and thus its features partly resemble a weak cation exchanger, but for Capto™ MMC also several other types of interactions are involved, including hydrogen bonding and hydrophobic interactions.
Capto™ MMC is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto™ MMC meets industrial demands with security of supply and comprehensive technical and regulatory support.
Features and Benefits
- Multimodal cation exchanger with a novel selectivity.
- High dynamic binding capacity at high conductivity increases productivity
- High-volume throughput cuts process times
- Smaller unit operations give cost-effective processing and reduce investments
Storage and Stability
Analysis Note
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Capto MMC is a multimodal cation exchanger with the properties of a weak cation exchanger. In addition to electrostatic interactions, the ligand structure provides for additional interaction modes such as hydrophobic interaction, hydrogen bonding, and thiophilic interaction.
This page describes the maintenance of media and storage conditions for multimodal chromatography using Cytiva products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service