Skip to Content
Merck
All Photos(2)

Documents

GBL162450

Grace Bio-Labs coverglass, 1oz

L × W × thickness 61 mm × 0.17 mm × 1.5 mm

Synonym(s):

glass slide, microscopy slide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.15

material

silicone (on glass)

sterility

non-sterile

packaging

pkg of 50 ea

manufacturer/tradename

Grace Bio-Labs 162450

L × W × thickness

61 mm × 0.17 mm × 1.5 mm

General description

A thin flat coverglass is the optimal platform for high resolution microscopy and high-sensitivity, quantitative, fluorescent imaging.

Chambered coverglass are used to isolate cells using removable hydrophobic barriers. CultureWell chambered coverglass leak-proof design eliminates well-to-well cross contamination. Products may be placed in sterile culture plates or 100mm diameter culture dishes. This product consists of the coverglass only, no gaskets included. Product is provided in 1 oz quantities.

Application

High resolution microscopy
Fluorescent imaging

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service