G9168
GR 218231
>98% (HPLC), solid
Synonym(s):
(+)-(2R)-1,2,3,4-Tetrahydro-6-[[(4-methoxyphenyl)sulfonyl]methyl]-N,N-dipropyl-2-naphthalenamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C24H33NO3S
CAS Number:
Molecular Weight:
415.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
>98% (HPLC)
form
solid
color
tan
solubility
DMSO: soluble ~24 mg/mL at 60 °C
SMILES string
CCCN(CCC)[C@@H]1CCc2cc(CS(=O)(=O)c3ccc(OC)cc3)ccc2C1
Biochem/physiol Actions
Selective D3 dopamine receptor antagonist.
Legal Information
Sold for research purposes under agreement with GlaxoSmithKline
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service