Skip to Content
Merck
All Photos(1)

Key Documents

G5761

Sigma-Aldrich

Glycine β-naphthylamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O · HCl
CAS Number:
Molecular Weight:
236.70
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

form

powder

technique(s)

protein purification: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

Cl.NCC(=O)Nc1ccc2ccccc2c1

InChI

1S/C12H12N2O.ClH/c13-8-12(15)14-11-6-5-9-3-1-2-4-10(9)7-11;/h1-7H,8,13H2,(H,14,15);1H

InChI key

CDOBGWIHUYXBJY-UHFFFAOYSA-N

Application

Glycine β-naphthylamide is used to identify, differentiate and characterize various amino acid β-naphthylamidase(s)/aminopeptidase(s).

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiyoshi Ito et al.
Bioscience, biotechnology, and biochemistry, 67(1), 83-88 (2003-03-07)
Glycyl aminopeptidase was purified 600-fold from a cell extract of Actinomucor elegans by ammonium sulfate fractionation and sequential chromatography on DEAE-Toyopearl, Toyopearl HW65C, and FPLC-Superdex 200 HR, with recovery of 3.3% of the activity. The enzyme highly specifically hydrolyzed Gly-X
P F Silveira et al.
Peptides, 22(12), 2137-2144 (2002-01-12)
Enzymatic cleavage of some peptides could be included among the mechanisms of water-electrolyte homeostasis. To test this hypothesis, the angiotensin-converting activity (ACE) of plasma and the L-cystine-di-beta-naphthylamidase activity (CAP) of plasma and of soluble and particulate fractions from different areas
Anna Musidlowska-Persson et al.
Protein engineering, 16(12), 1139-1145 (2004-02-26)
It was shown recently that proline-beta-naphthylamidase from pig liver resembles the gamma-subunit of pig liver esterase (PLE), which could be functionally expressed in the yeast Pichia pastoris in recombinant form (rPLE). The gene encoding rPLE shares 97% identity with the
Endang Setyorini et al.
Journal of basic microbiology, 46(4), 294-304 (2006-07-19)
A halotolerant strain FP-133, able to grow at concentrations of 0-12.5% (w/v) NaCl, was isolated from a fish paste and identified as Bacillus subtilis . B. subtilis strain FP-133 produced an intracellular protease which showed catalytic activity under saline conditions.
T Takahashi et al.
The Journal of biological chemistry, 264(20), 11565-11571 (1989-07-15)
An enzyme hydrolyzing proline-beta-naphthylamide was purified to apparent homogeneity from porcine intestinal mucosa. The purified enzyme appears to consist of three identical subunit polypeptides with a molecular weight of about 58,000 each, associated noncovalently. The enzyme is a glycoprotein, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service