G4274
Globoside from human erythrocytes
≥95%
Synonym(s):
Globotetraosylceramide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human erythrocytes
Assay
≥95%
form
film
functional group
amide
hydroxyl
shipped in
ambient
storage temp.
−20°C
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
International journal of molecular sciences, 21(10) (2020-05-28)
Glycosphingolipids containing very-long-chain fatty acids (VLCFAs) regulate several immune responses, such as cytokine production, immune signaling, and antibody induction. We previously reported that stimulation with an inflammatory mediator, TNF-α, promotes the expression of glycosphingolipids in vascular endothelial cells. The major
The Journal of biological chemistry, 273(5), 2517-2525 (1998-02-28)
Undifferentiated human embryonal carcinoma cells are characterized by high expression of lactoneotetraosylceramide (nLc4), globoside (Gb4), and extended globo-series glycosphingolipids (GSLs) termed "stage-specific embryonic antigens 3 and 4" (SSEA-3 and -4). Expression of these GSLs declines in association with a decline
Biological chemistry, 393(3), 133-147 (2012-06-22)
Membrane microdomain association of the glycosphingolipids (GSLs) globotriaosylceramide (Gb3Cer) and globotetraosylceramide (Gb4Cer), the highly and less effective receptors, respectively, for Shiga toxins (Stxs), is assumed as a functional requirement for Stx-mediated cytotoxicity. In a previous study, we demonstrated predominant localization
Biochemistry, 36(28), 8539-8547 (1997-07-15)
Recognition of macromolecules by glycosphingolipids is closely correlated with the nature of the glycolipid carbohydrate; however, it is also thought to be secondarily modulated by the structure of the single fatty acid. In the present work, we sought insight into
Journal of virology, 84(22), 11737-11746 (2010-09-10)
Globoside (Gb4Cer), Ku80 autoantigen, and α5β1 integrin have been identified as cell receptors/coreceptors for human parvovirus B19 (B19V), but their role and mechanism of interaction with the virus are largely unknown. In UT7/Epo cells, expression of Gb4Cer and CD49e (integrin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service