Skip to Content
Merck
All Photos(1)

Key Documents

F9142

Supelco

Flow adapter for jacketed LC columns

column I.D. 2.5 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.52

technique(s)

LPLC: suitable

column I.D.

2.5 cm

Looking for similar products? Visit Product Comparison Guide

General description

Each flow adapter is supplied with a 20 μm porosity HDPE bed support, TFE/polypropylene O-ring seal, stainless steel adjusting rod, PTFE body and 5 feet (1.5 meters) of FEP tubing.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service