Skip to Content
Merck
All Photos(1)

Key Documents

F4761

Sigma-Aldrich

Formamide

ACS reagent, ≥99.5% (GC)

Synonym(s):

Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

ACS reagent

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

Assay

≥99.5% (GC)

autoignition temp.

932 °F

color

APHA: ≤10

refractive index

n20/D 1.447 (lit.)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service