Skip to Content
Merck
All Photos(1)

Key Documents

F4261

Sigma-Aldrich

α-L-Fucopyranosylphenyl isothiocyanate

suitable for preparation of neoglycoproteins

Synonym(s):

Isothiocyanatophenyl α-L-fucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO5S
CAS Number:
Molecular Weight:
297.33
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

suitability

suitable for preparation of neoglycoproteins

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Monsigny et al.
Biology of the cell, 51(2), 187-196 (1984-01-01)
The presence and the sugar specificity of membrane lectins on the cell surface of mouse L1210 leukemia cells were demonstrated by using various neoglycoproteins (glycosylated serum albumin) substituted with fluorescein or methotrexate. Neoglycoproteins were prepared by reaction of glycosidophenylisothiocyanates with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service