Skip to Content
Merck
All Photos(1)

Key Documents

F0176

Sigma-Aldrich

Anti-FLI-1 antibody produced in sheep

fractionated antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

sheep

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 52 kDa

species reactivity

human, mouse

technique(s)

western blot: suitable using whole extracts from mouse erythroleukemia cell lines, DP16-1 and CB7 and human T-cell line.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FLI1(2313)
mouse ... Fli1(14247)

Immunogen

GST fusion protein of the N-terminus of Fli-1 corresponding to amino acids 1-239.

Application

Anti-FLI-1 antibody produced in sheep is suitable for western blotting using whole extracts from mouse erythroleukemia cell lines, DP16-1 and CB7 and human T-cell line.

Biochem/physiol Actions

Friend leukemia integration 1 transcription factor (FLI1) is a protein that is encoded by the FLI1 gene in humans and is a member of Ets transcription factor family. It is a proto-oncogene and located on human chromosome 11q23-24. FLI1 may be involved in human sarcomas, leukemias and lymphomas. It may play an important role in the posttranscriptional regulation of the Fli-1 transcript. Alteration in the transcriptional activation function of Fli-1 may be responsible for human malignancies. It is a novel ETS transcription factor involved in gene fusions in prostate cancer.

Physical form

Solution in 0.01 M phosphate buffered saline, containing 0.08% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V N Rao et al.
Oncogene, 8(8), 2167-2173 (1993-08-01)
Three of the ets oncogene superfamily members v-ets, Spi-1/PU.1 and Fli-1, have been shown to be directly involved in retroviral-mediated acute erythroleukemias. The Fli-1 gene was found to be rearranged in 75% of the erythroleukemias induced by Friend murine leukemia
Steven M Kornblau et al.
Blood, 118(20), 5604-5612 (2011-09-16)
Friend leukemia virus integration 1 (FLI1), an Ets transcription factor family member, is linked to acute myelogenous leukemia (AML) by chromosomal events at the FLI1 locus, but the biologic impact of FLI1 expression on AML is unknown. FLI1 protein expression
Paula Paulo et al.
Genes, chromosomes & cancer, 51(3), 240-249 (2011-11-15)
To characterize the pattern of ETS rearrangements and to uncover novel ETS fusion genes, we analyzed 200 prostate carcinomas (PCa) with TaqMan low-density arrays (TLDAs), followed by selective analyses with fluorescence in situ hybridization (FISH), RT-PCR, and sequencing. Besides confirming
D D Prasad et al.
Cancer research, 52(20), 5833-5837 (1992-10-15)
Three ets family members, v-ets, spleen focus forming virus proviral integration 1/Pu.1, and Friend leukemia integration 1 (Fli-1), were shown to be involved in retroviral mediated acute leukemias suggesting that ets family members play a crucial role in transformation. Mouse
V Baud et al.
Genomics, 11(1), 223-224 (1991-09-01)
FLI1 is a common mouse viral integration region in virus-induced leukemias and lymphomas. Using an evolutionarily conserved mouse probe and Southern hybridization to (rodent x human) somatic cell hybrid DNAs, the human homolog of FLI1 has been shown to lie

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service