EP6001040110
Eppendorf® Chest Freezer Rack
Holds 14 x DWP (53 mm/2 in), (14 x 1) configuration, stainless steel, side access
About This Item
Recommended Products
material
stainless steel
feature
(14 x 1) configuration
Holds 14 x DWP (53 mm/2 in)
max. DWP number per rack: 14 (14 x 1)
side access
manufacturer/tradename
Eppendorf® 6001040110 (Global)
technique(s)
cryopreservation: suitable
D × W × H
133 mm (5.2 in.) × 91 mm (3.6 in.) × 776 mm (30.6 in.)
capacity
(max box size (mm): 86 x 128)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service