Key Documents
EP5810722004
Eppendorf® Adapter for 500 mL Rectangular Bucket
holds 12x15 mL conical tubes, pack of 2 ea
Select a Size
2 EA
1.590,00 kr.
1.590,00 kr.
Please contact Customer Service for Availability
Select a Size
Change View
2 EA
1.590,00 kr.
About This Item
UNSPSC Code:
41103911
NACRES:
NB.78
1.590,00 kr.
Please contact Customer Service for Availability
feature
holds 12x15 mL conical tubes
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5810722004
technique(s)
centrifugation: suitable
compatibility
for use with 500 mL rectangular buckets
General description
Adapter for use with Eppendorf 500 mL rectangular bucket.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)