Documents
EP5702740004
Eppendorf® Swing-bucket rotor A-4-38
without buckets
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
without buckets (without buckets)
without buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5702740004
parameter
4,400 max. RCF (2,750 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5702/5702R/5702RH
Legal Information
Eppendorf is a registered trademark of Eppendorf AG