EP5427736000
Eppendorf® Adapter
holds 1 x 20-30 mL, bore size (large), pack of 2 ea
Select a Size
640,00 kr.
Select a Size
About This Item
640,00 kr.
feature
holds 1 x 20-30 mL
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5427736000
technique(s)
centrifugation: suitable
bore size
(large)
compatibility
for use with Rotor F-35-6-30 (large rotor bore)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)