EP5420000245
Eppendorf® Centrifuge 5420
keypad, refrigerated: no, without rotor, AC/DC input 120 V, 50 - 60 Hz
Synonym(s):
Centrifuge 5420 - Microcentrifuge, Eppendorf® 120 V Centrifuge 5420
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
CE compliant
CSA approved
refrigerated: no
keypad
without rotor
Application
Centrifugation
Features and Benefits
with keypad control panel for easy cleaningwithout rotornon-refrigerated120 V, 50 – 60 Hz (US)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service