Documents
EP5353040156
Eppendorf® Tube Holder
25/50 mL
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
feature
25/50 mL
packaging
case of 1 ea
manufacturer/tradename
Eppendorf® 5353040156
Related Categories
General description
  • High-speed mixer (3,000 rpm) for fast and efficient mixing of small volumes in all common vessel formats up to 50 mL
  • 2DMix-Control technology (planar mixing orbit without vertical movement) for controlled mixing and reproducible results in seconds
  • Anti-spill technology for controlled mixing movements without lid wetting and cross-contamination
  • 5 different tube holders (0.5 mL, 1.5/2.0 mL, 5/15 mL, 30/50 mL, PCR 96-well plates) for high vessel flexibility
  • Integrated ergonomic vortex function for various vessel formats for distinct sample mixing
  • Minimal hand/arm vibration transfer during vortexing (certificate available) for ergonomic operation
  • 5 soft keys with pre-defined standard mixing parameters for easy usage
  • Very stable - remains stationary even at maximum mixing speed
  • Whisper quiet operation to benefit the work environment
Legal Information
Eppendorf is a registered trademark of Eppendorf AG