EP4924000940
Eppendorf® Reference 2 Pipettes with Carousel and epT.I.P.S.®
Eppendorf Reference 2 Pipettes with Carousel and epT.I.P.S.
Synonym(s):
pipettor holder
Select a Size
14.600,00 kr.
Select a Size
About This Item
14.600,00 kr.
Recommended Products
feature
no. of channels 1 x Channel
Mechanical
packaging
pack of 1
manufacturer/tradename
Eppendorf® 4924000940
volume
0.1- 2.5 μL
0.5- 10 μL
100 - 1,000 μL
10 - 100 μL
20 - 200 μL
2 - 20 μL
Related Categories
General description
Application
Features and Benefits
- Mechanical air-cushion pipette with outstanding precision and accuracy - operated with one button
- With its innovative one-button operation, the Reference 2 allows fast and ergonomic handling with reduced operating efforts and active aerosol reduction. A clear haptic feedback helps to avoid accidental tip ejection
- Spring-loaded tip cones improve user-to-user reproducibility and ergonomics by lowering the tip attachment force
- The unique smooth surface and full autoclavability guarantee efficient decontamination - making it the ideal companion when working under sterile conditions
- A temporary adjustment allows for an easy adjustment of your Reference 2 for the most accurate pipetting of different liquids or in changing external conditions without the need for a full calibration
- Available as single-channel pipette in fixed or variable volume as well as 8- and 12-channel pipette
- The Reference 2 multi-channel variants offer high flexibility incl. a spring-loaded tip cone which can be switched on/off optionally
Packaging
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service