Key Documents
EP022822144
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-45-24-12, non aerosol tight, (24 x 6.0/8.0 mL round-bottom tubes)
About This Item
UNSPSC Code:
41103909
Product EP022822144 is not currently sold in your country. Contact Technical Service
feature
to hold 24 x 12x67 - 100 mm tubes
to hold 24 x 6.0/8.0 mL round-bottom tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022822144 (NA)
technique(s)
centrifugation: suitable
capacity
(24 x 6.0/8.0 mL round-bottom tubes)
compatibility
for use with centrifuge models Vacufuge Plus Concentrator
General description
Selection of rotors for the Eppendorf Vacufuge Plus.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)