EP022638935
Buckets for Eppendorf® S-4-104 Rotor
plate bucket, aerosol-tight capable, with plate carrier, pack of 4
Synonym(s):
Rotor buckets
About This Item
Recommended Products
feature
aerosol-tight capable
plate bucket
with plate carrier
packaging
pack of 4
manufacturer/tradename
Eppendorf® 022638935 (NA)
technique(s)
centrifugation: suitable
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Plate buckets have a maximum speed of 2,568 × g (3,900 rpm)
- Package includes 4 plate buckets with a plate carrier
- Autoclavable at 121 °C for 20 min
- Buckets feature aerosol-tight capable caps for protection against hazardous materials during centrifugation
- Compatible with rotor S-4-104 for 5810/5810R centrifuge
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service