EP0030128613
Eppendorf® Twin tec PCR plates, 96 wells
red plate, conical bottom colorless wells, semi-skirted, pkg of 25 ea
Select a Size
1.150,00 kr.
Select a Size
About This Item
polycarbonate
red plate
1.150,00 kr.
Recommended Products
material
conical bottom colorless wells
polycarbonate
red plate
sterility
non-sterile
feature
skirt (Semi skirted)
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128613
technique(s)
PCR: suitable
size
96 wells
well maximum volume
250 μm
General description
Features and Benefits
- Eppendorf® Twin tec PCR plate combine a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- Cut-away corner and alphanumeric grid-referencing.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
- Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors.
- Fits most thermal cyclers.
- 250 μL max. well volume (when used with cap strips).
- Improved well-to-well tolerance.
- Ideal for quantitative real-time PCR as well – OptiTrack® matrix for faster sample identification and fewer pipetting errors.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)